Kalium(4-(benzyloxy)phenyl)trifluoroborat

Lieferant: Apollo Scientific

Synonyme: Potassium (4-(benzyloxy)phenyl)trifluoroborate

PC11238-5G
APOSPC11238-5GEA 115 EUR
APOSPC11238-5G
Kalium(4-(benzyloxy)phenyl)trifluoroborat
Kalium(4-(benzyloxy)phenyl)trifluoroborat
Formel: C₁₃H₁₁BF₃KO
Molecular Weight: 290,13 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115746
CAS-Nummer: 850623-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR