Kalium(3-methoxyphenyl)trifluoroborat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Methoxyphenyl)trifluorborsäure Kaliumsalz
PC11243-1G
PC11243-250MG
PC11243-5G
APOSPC11243-1GEA
58.2
EUR
APOSPC11243-1G
APOSPC11243-250MG
APOSPC11243-5G
Kalium(3-methoxyphenyl)trifluoroborat
Kalium(3-methoxyphenyl)trifluoroborat
Formel:
C₇H₇BF₃KO Molecular Weight: 214,04 g/mol Schmelzpunkt: 164…168 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039954 CAS-Nummer: 438553-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...