Kalium(4-(methylsulfonyl)phenyl)trifluoroborat
Lieferant: Apollo Scientific
Synonyme:
Potassium trifluoro(4-(methylsulphonyl)phenyl)borate
PC11244-1G
PC11244-5G
APOSPC11244-1GEA
105
EUR
APOSPC11244-1G
APOSPC11244-5G
Kalium(4-(methylsulfonyl)phenyl)trifluoroborat
Potassium-4-(methylsulphonyl)phenyltrifluoroborate
Formel:
C₇H₇BF₃KO₂S Molecular Weight: 262,09 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115738 CAS-Nummer: 850623-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...