3-(4-Fluorphenylcarbamoyl)phenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC11247-1G
PC11247-5G
APOSPC11247-1GEA
170
EUR
APOSPC11247-1G
APOSPC11247-5G
3-(4-Fluorphenylcarbamoyl)phenylboronsäure 97%
3-(4-Fluorphenylcarbamoyl)phenylboronsäure
Formel:
C₁₃H₁₁BFNO₃ Schmelzpunkt: 232…238 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04115712 CAS-Nummer: 850567-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...