Pentafluorophenyl-8-chinolinsulfonat

Lieferant: Apollo Scientific

PC11270-5G PC11270-1G PC11270-500MG
APOSPC11270-5GEA 759 EUR
APOSPC11270-5G APOSPC11270-1G APOSPC11270-500MG
Pentafluorophenyl-8-chinolinsulfonat
Pentafluorophenyl-8-chinolinsulfonat
Formel: C₉H₇NO₃S
Schmelzpunkt: 155…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05975139
CAS-Nummer: 885950-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR