Pentafluorphenyl-4-(acetylamino)benzolsulfonat

Lieferant: Apollo Scientific

PC11272-1G PC11272-500MG PC11272-5G
APOSPC11272-1GEA 275 EUR
APOSPC11272-1G APOSPC11272-500MG APOSPC11272-5G
Pentafluorphenyl-4-(acetylamino)benzolsulfonat
Pentafluorphenyl-4-(acetylamino)benzolsulfonat
Formel: C₁₄H₈F₅NO₄S
Schmelzpunkt: 168…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05975141
CAS-Nummer: 886360-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR