3,5-Difluorsalicylsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Difluoro-2-hydroxybenzoic acid

PC1142-1G PC1142-5G
APOSPC1142-1GEA 87 EUR
APOSPC1142-1G APOSPC1142-5G
3,5-Difluorsalicylsäure
3,5-Difluorsalicylsäure
Formel: C₇H₄F₂O₃
Molecular Weight: 174.10 g/mol
Schmelzpunkt: 199…203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06203609
CAS-Nummer: 84376-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR