2-Brom-2'-chlor-5'-fluoracetophenon 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Chloro-5-fluorophenacyl bromide

PC1153-1G PC1153-5G
APOSPC1153-1GEA 65.5 EUR
APOSPC1153-1G APOSPC1153-5G
2-Brom-2'-chlor-5'-fluoracetophenon 98%
2-Brom-2'-chlor-5'-fluoracetophenon
Formel: C₈H₅BrClFO
Lagertemperatur: Kühlschrank
MDL: MFCD08059509
CAS-Nummer: 76609-34-2
UN: 1760
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR