6-Fluoro-1H-indole-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Fluoroindole-2-carboxylic acid
PC1239-1G
PC1239-5G
APOSPC1239-1GEA
21.9
EUR
APOSPC1239-1G
APOSPC1239-5G
6-Fluoro-1H-indole-2-carboxylic acid
6-Fluoro-1H-indole-2-carboxylic acid
Formel:
C₉H₆FNO₂ Molecular Weight: 179,15 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01863162 CAS-Nummer: 3093-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...