Gallium(III) trifluoromethanesulphonate

Lieferant: Apollo Scientific
Gefahr

Synonyme: Gallium tris(trifluoromethanesulphonate)

PC1246-10G PC1246-25G PC1246-5G
APOSPC1246-10GEA 139 EUR
APOSPC1246-10G APOSPC1246-25G APOSPC1246-5G
Gallium(III) trifluoromethanesulphonate
Gallium(III) trifluoromethanesulphonate
Formel: C₃F₉GaO₉S₃
Molecular Weight: 516,913 g/mol
Dichte: 1,7 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD02093898
CAS-Nummer: 74974-60-0
UN: 3260
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR