2-Bromo-3,3,4,4,4-pentafluoro-1-butene 98%

Lieferant: Apollo Scientific

PC1265-1G PC1265-5G
APOSPC1265-1GEA 120 EUR
APOSPC1265-1G APOSPC1265-5G
2-Bromo-3,3,4,4,4-pentafluoro-1-butene 98%
2-Bromo-3,3,4,4,4-pentafluoro-1-butene
Formel: C₄H₂BrF₅
Molecular Weight: 224,96 g/mol
Siedepunkt: 56…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094107
CAS-Nummer: 68318-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR