n-Butyl 2,2,2-trifluoroethylacetaldehyde acetal 95%
Lieferant: Apollo Scientific
Synonyme:
Acetaldehyde butyl 2,2,2-trifluoroethyl acetal
PC1267-100G
PC1267-25G
APOSPC1267-100GEA
911
EUR
APOSPC1267-100G
APOSPC1267-25G
n-Butyl 2,2,2-trifluoroethylacetaldehyde acetal 95%
n-Butyl 2,2,2-trifluoroethylacetaldehyde acetal
Formel:
C₈H₁₅F₃O₂ Molecular Weight: 200.2 g/mol Siedepunkt: 143.2…143.3 °C/751mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094109 CAS-Nummer: 2925-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...