3-Chlor-1-fluor-4-nitrobenzol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chlor-4-fluornitrobenzol

PC1336-1G PC1336-25G PC1336-5G
APOSPC1336-1GEA 21.9 EUR
APOSPC1336-1G APOSPC1336-25G APOSPC1336-5G
3-Chlor-1-fluor-4-nitrobenzol
3-Chlor-1-fluor-4-nitrobenzol
Formel: C₆H₃ClFNO₂
Molecular Weight: 175,55 g/mol
Schmelzpunkt: 33…37 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03412200
CAS-Nummer: 2106-50-5
EINECS: 218-286-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR