2-Fluor-4-methoxybenzonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluor-4-methoxybenzoesäurenitril

PC1358-10G PC1358-5G PC1358-25G
APOSPC1358-10GEA 49.3 EUR
APOSPC1358-10G APOSPC1358-5G APOSPC1358-25G
2-Fluor-4-methoxybenzonitril 98%
2-Fluor-4-methoxybenzonitril
Formel: C₈H₆FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00509374
CAS-Nummer: 94610-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR