1-Bromo-4-chloro-1,1,2,2-tetrafluorobutane 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1369B-25G
PC1369B-5G
PC1369B-1G
APOSPC1369B-25GEA
0
EUR
APOSPC1369B-25G
APOSPC1369B-5G
APOSPC1369B-1G
1-Bromo-4-chloro-1,1,2,2-tetrafluorobutane 97%
1-Bromo-4-chloro-1,1,2,2-tetrafluorobutane
Formel:
C₄H₄BrClF₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153677 CAS-Nummer: 232602-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...