3-Bromo-2-fluorophenylacetic acid

Lieferant: Apollo Scientific
Gefahr

PC1378-1G PC1378-5G PC1378-25G
APOSPC1378-1GEA 74.1 EUR
APOSPC1378-1G APOSPC1378-5G APOSPC1378-25G
3-Bromo-2-fluorophenylacetic acid
3-Bromo-2-fluorophenylacetic acid
Formel: C8H6BrFO2
Molecular Weight: 233,0300 g/mol
Schmelzpunkt: 133…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09864663
CAS-Nummer: 1375964-39-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR