2-Fluor-m-toluonitril
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Fluoro-3-methylbenzonitrile
PC1380-25G
PC1380-5G
APOSPC1380-25GEA
81.2
EUR
APOSPC1380-25G
APOSPC1380-5G
2-Fluor-m-toluonitril
2-Fluor-m-toluonitril
Formel:
C₈H₆FN Molecular Weight: 135,1384 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07782078 CAS-Nummer: 185147-07-3 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...