2'-Brom-4'-fluoracetophenon 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Bromo-4-fluorophenyl)ethanone

PC1396B-25G PC1396B-5G PC1396B-100G
APOSPC1396B-25GEA 101 EUR
APOSPC1396B-25G APOSPC1396B-5G APOSPC1396B-100G
2'-Brom-4'-fluoracetophenon 97%
2'-Brom-4'-fluoracetophenon
Formel: C₈H₆BrFO
Molecular Weight: 217.04 g/mol
Siedepunkt: 150 °C/12mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00077464
CAS-Nummer: 1006-39-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR