2-Brom-α,α,α,3-tetrafluortoluol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Brom-3-fluorbenzotrifluorid, 1-Brom-2-fluor-6-(trifluormethyl)benzol
PC1420T-1G
PC1420T-25G
PC1420T-5G
APOSPC1420T-1GEA
21.9
EUR
APOSPC1420T-1G
APOSPC1420T-25G
APOSPC1420T-5G
2-Brom-α,α,α,3-tetrafluortoluol 98%
2-Brom-α,α,α,3-tetrafluortoluol
Formel:
C₇H₃BrF₄ Molecular Weight: 243 g/mol Dichte: 1,741 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061243 CAS-Nummer: 104540-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...