4-Brom-α,α,α,3-tetrafluortoluol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1423A-25G
PC1423A-5G
PC1423A-100G
APOSPC1423A-25GEA
36.4
EUR
APOSPC1423A-25G
APOSPC1423A-5G
APOSPC1423A-100G
4-Brom-α,α,α,3-tetrafluortoluol 97%
4-Brom-α,α,α,3-tetrafluortoluol
Formel:
C₇H₃BrF₄ Molecular Weight: 243 g/mol Siedepunkt: 154…155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042497 CAS-Nummer: 40161-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...