4-Brom-2-fluorbenzylamin Hydrochlorid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Bromo-2-fluorophenyl)methanamine hydrochloride

PC1423G-25G PC1423G-5G
APOSPC1423G-25GEA 92.8 EUR
APOSPC1423G-25G APOSPC1423G-5G
4-Brom-2-fluorbenzylamin Hydrochlorid 98%
4-Brom-2-fluorbenzylamin Hydrochlorid
Formel: C₇H₇BrFN·HCl
Molecular Weight: 240.50 g/mol
Schmelzpunkt: 254…257 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143266
CAS-Nummer: 147181-08-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR