4-Brom-2-fluorzimtsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(4-Brom-2-fluorphenyl)propensäure, 4-Brom-2-fluorcinnamsäure
PC1423P-25G
PC1423P-5G
PC1423P-100G
APOSPC1423P-25GEA
46.4
EUR
APOSPC1423P-25G
APOSPC1423P-5G
APOSPC1423P-100G
4-Brom-2-fluorzimtsäure 98%
4-Brom-2-fluorzimtsäure
Formel:
C₉H₆BrFO₂ Molecular Weight: 245.05 g/mol Siedepunkt: 322 Schmelzpunkt: 216…221 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143267 CAS-Nummer: 149947-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...