Ethyl-2-chloro-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

PC14280-100MG PC14280-250MG
APOSPC14280-100MGEA 21.9 EUR
APOSPC14280-100MG APOSPC14280-250MG
Ethyl-2-chloro-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate
Ethyl-2-chloro-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate
Formel: C₇H₅ClF₃NO₂S
Molecular Weight: 259,64 g/mol
Schmelzpunkt: 57…59 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02956778
CAS-Nummer: 72850-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR