3-Chloro-2-ethyl-5-(trifluoromethyl)pyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC1442-1G
PC1442-500MG
APOSPC1442-1GEA
119
EUR
APOSPC1442-1G
APOSPC1442-500MG
3-Chloro-2-ethyl-5-(trifluoromethyl)pyridine 95%
3-Chloro-2-ethyl-5-(trifluoromethyl)pyridine
Formel:
C₈H₇ClF₃N Siedepunkt: 60 °C/15mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00202365 CAS-Nummer: 303147-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...