1-(2-Fluorophenyl)-1H-pyrrole-2-carbaldehyde 95%

Lieferant: Apollo Scientific

PC1446-1G PC1446-25G PC1446-5G
APOSPC1446-1GEA 133 EUR
APOSPC1446-1G APOSPC1446-25G APOSPC1446-5G
1-(2-Fluorophenyl)-1H-pyrrole-2-carbaldehyde 95%
1-(2-Fluorophenyl)-1H-pyrrole-2-carbaldehyde
Formel: C₁₁H₈FNO
Molecular Weight: 189,19 g/mol
Schmelzpunkt: 123…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01443629
CAS-Nummer: 132407-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR