3-Chlor-2,6-difluorbenzoesäure 95%

Lieferant: Apollo Scientific
Achtung

PC1457-100G PC1457-10G PC1457-50G
APOSPC1457-100GEA 377 EUR
APOSPC1457-100G APOSPC1457-10G APOSPC1457-50G
3-Chlor-2,6-difluorbenzoesäure 95%
3-Chloro-2,6-difluorobenzoic acid
Formel: C₇H₃ClF₂O₂
Molecular Weight: 192.55 g/mol
Schmelzpunkt: 126…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631322
CAS-Nummer: 225104-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR