2-Bromo-3,4,4,5,5,5-hexafluoro-3-(trifluoromethyl)pent-1-ene 97%

Lieferant: Apollo Scientific
Achtung

PC1472-25G PC1472-5G
APOSPC1472-25GEA 0 EUR
APOSPC1472-25G APOSPC1472-5G
2-Bromo-3,4,4,5,5,5-hexafluoro-3-(trifluoromethyl)pent-1-ene 97%
2-Bromo-3,4,4,5,5,5-hexafluoro-3-(trifluoromethyl)pent-1-ene
Formel: C₆H₂BrF₉
Lagertemperatur: Raumtemperatur
MDL: MFCD00153704
CAS-Nummer: 234096-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR