2-Brom-α,α,α-trifluor-5-nitrotoluol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Brom-5-nitrobenzotrifluorid, 1-Brom-4-nitro-2-(trifluormethyl)benzol
PC1490-25G
PC1490-100G
APOSPC1490-25GEA
21.9
EUR
APOSPC1490-25G
APOSPC1490-100G
2-Brom-α,α,α-trifluor-5-nitrotoluol 98%
2-Brom-α,α,α-trifluor-5-nitrotoluol
Formel:
C₇H₃BrF₃NO₂ Molecular Weight: 270,01 g/mol Siedepunkt: 87…88 °C/3mmHg Schmelzpunkt: 41…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014707 CAS-Nummer: 367-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...