5-Fluor-2-picolin Hydrochlorid 98+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Fluoro-2-methylpyridine
PC1509-1G
PC1509-5G
PC1509-25G
APOSPC1509-1GEA
39.3
EUR
APOSPC1509-1G
APOSPC1509-5G
APOSPC1509-25G
5-Fluor-2-picolin Hydrochlorid 98+%
5-Fluor-2-picolin Hydrochlorid
Formel:
C₆H₇ClFN Molecular Weight: 111,1169 g/mol Siedepunkt: 114,7 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095271 CAS-Nummer: 31181-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...