4-Brom-2,6-difluorbenzonitril

Lieferant: Apollo Scientific
Gefahr

PC1515-25G PC1515-5G PC1515-100G
APOSPC1515-25GEA 87 EUR
APOSPC1515-25G APOSPC1515-5G APOSPC1515-100G
4-Brom-2,6-difluorbenzonitril
4-Brom-2,6-difluorbenzonitril
Formel: C₇H₂BrF₂N
Molecular Weight: 218 g/mol
Schmelzpunkt: 79…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01310981
CAS-Nummer: 123843-67-4
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR