2-Chlor-3-fluorphenylboronsäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-3-fluorobenzeneboronic acid, (2-Chloro-3-fluorophenyl)boronic acid
PC1554-1G
PC1554-250MG
APOSPC1554-1GEA
75.4
EUR
APOSPC1554-1G
APOSPC1554-250MG
2-Chlor-3-fluorphenylboronsäure 99%
2-Chlor-3-fluorphenylboronsäure
Formel:
C₆H₅BClFO₂ Molecular Weight: 174,37 g/mol Schmelzpunkt: 80…84 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07363741 CAS-Nummer: 871329-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...