5-Brom-2-(trifluormethoxy)anilin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Amino-4-(trifluoromethoxy)bromobenzene
PC1556-100G
PC1556-25G
PC1556-5G
APOSPC1556-100GEA
97.3
EUR
APOSPC1556-100G
APOSPC1556-25G
APOSPC1556-5G
5-Brom-2-(trifluormethoxy)anilin 98%
5-Brom-2-(trifluormethoxy)anilin
Formel:
C₇H₅BrF₃NO Molecular Weight: 256.0200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039242 CAS-Nummer: 886762-08-9 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...