4-Brom-2-nitro(trifluormethoxy)benzol

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-(trifluoromethoxy)nitrobenzene, 3-Nitro-4-(trifluoromethoxy)bromobenzene

PC1557-25G PC1557-5G PC1557-100G
APOSPC1557-25GEA 24.8 EUR
APOSPC1557-25G APOSPC1557-5G APOSPC1557-100G
4-Brom-2-nitro(trifluormethoxy)benzol
4-Brom-2-nitro(trifluormethoxy)benzol
Formel: C₇H₃BrF₃NO₃
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039243
CAS-Nummer: 95668-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR