2-Amino-5-brom-2'-fluorbenzophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Amino-5-bromophenyl)(2-fluorophenyl)methanone, 2-Amino-2'-fluoro-5-bromobenzophenone

PC1566-25G PC1566-5G
APOSPC1566-25GEA 191 EUR
APOSPC1566-25G APOSPC1566-5G
2-Amino-5-brom-2'-fluorbenzophenon
2-Amino-5-brom-2'-fluorbenzophenon
Formel: C₁₃H₉BrFNO
Molecular Weight: 294.12 g/mol
Schmelzpunkt: 100…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038380
CAS-Nummer: 1479-58-9
EINECS: 216-037-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR