3-Fluorthioanisol

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Fluor-3-(methylthio)benzol, 3-Fluorphenylmethylsulfid

PC1579-100G PC1579-25G PC1579-5G
APOSPC1579-100GEA 696 EUR
APOSPC1579-100G APOSPC1579-25G APOSPC1579-5G
3-Fluorthioanisol
3-Fluorthioanisol
Formel: C₇H₇FS
Molecular Weight: 142.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03789131
CAS-Nummer: 658-28-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR