6-Fluorchromon-2-carbonsäure 98%

Lieferant: Apollo Scientific

Synonyme: 6-Fluoro-4-oxo-4H-chromene-2-carboxylic acid

PC1589-1G PC1589-25G PC1589-5G
APOSPC1589-1GEA 109 EUR
APOSPC1589-1G APOSPC1589-25G APOSPC1589-5G
6-Fluorchromon-2-carbonsäure 98%
6-Fluorchromon-2-carbonsäure
Formel: C₁₀H₅FO₄
Molecular Weight: 208,15 g/mol
Schmelzpunkt: 257…259 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03070543
CAS-Nummer: 99199-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR