4'-Brom-2'-(trifluormethoxy)acetanilid 97%

Lieferant: Apollo Scientific

PC1592K-10G PC1592K-25G PC1592K-5G
APOSPC1592K-10GEA 135 EUR
APOSPC1592K-10G APOSPC1592K-25G APOSPC1592K-5G
4'-Brom-2'-(trifluormethoxy)acetanilid 97%
4'-Brom-2'-(trifluormethoxy)acetanilid
Formel: C₉H₇BrF₃NO₂
Schmelzpunkt: 135…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00203979
CAS-Nummer: 175278-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR