Z-L-Lys(Tfa)-OH 95%

Lieferant: Apollo Scientific

Synonyme: Z-Lys(Tfa)-OH

PC1597-25G PC1597-5G
APOSPC1597-25GEA 120 EUR
APOSPC1597-25G APOSPC1597-5G
Z-L-Lys(Tfa)-OH 95%
Z-L-Lys(Tfa)-OH
Formel: C₁₆H₁₉N₂O₅F₃
Molecular Weight: 376,31 g/mol
Schmelzpunkt: 88…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00270524
CAS-Nummer: 14905-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR