2-Brom-3,3,3-trifluorpropen 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-3,3,3-trifluoro-1-propene

PC1602-100G PC1602-25G PC1602-5G
APOSPC1602-100GEA 157 EUR
APOSPC1602-100G APOSPC1602-25G APOSPC1602-5G
2-Brom-3,3,3-trifluorpropen 98%
2-Bromo-3,3,3-trifluoropropene

Versatile CF3 building block: Hu et al, J. Chem Soc., Perkin Trans. 1, 1998, 2, 279. Precursor for PC77612-(Trifluoromethyl)acrylic acid : JOC., 1989 , 54.

Formel: C₃H₂BrF₃
Siedepunkt: 29…30 °C
Dichte: 1,66 g/cm³ (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00077469
CAS-Nummer: 1514-82-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR