3-Fluoro-5-hydroxybenzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-5-hydroxybenzeneboronic acid, 3-Fluoro-5-hydroxyphenylboronic acid, (3-Fluoro-5-hydroxyphenyl)boronic acid

PC1607-1G PC1607-250MG
APOSPC1607-1GEA 48 EUR
APOSPC1607-1G APOSPC1607-250MG
3-Fluoro-5-hydroxybenzeneboronic acid 98%
3-Fluoro-5-hydroxybenzeneboronic acid
Formel: C₆H₆BFO₃
Molecular Weight: 155,92 g/mol
Schmelzpunkt: 54…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363778
CAS-Nummer: 871329-82-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR