5-Chlor-2-(trifluormethyl)benzamid
Lieferant: Apollo Scientific
|
Achtung
|
PC1613-1G
PC1613-5G
APOSPC1613-1GEA
46.4
EUR
APOSPC1613-1G
APOSPC1613-5G
5-Chlor-2-(trifluormethyl)benzamid
5-Chlor-2-(trifluormethyl)benzamid
Formel:
C₈H₅ClF₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631406 CAS-Nummer: 654-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...