Methyl-2-(trifluoromethyl)pyridine-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 6-(trifluoromethyl)nicotinate
PC1616-1G
PC1616-5G
APOSPC1616-1GEA
21.9
EUR
APOSPC1616-1G
APOSPC1616-5G
Methyl-2-(trifluoromethyl)pyridine-5-carboxylate
Methyl-2-(trifluoromethyl)pyridine-5-carboxylate
Formel:
C₈H₆F₃NO₂ Molecular Weight: 205,13 g/mol Schmelzpunkt: 54…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001346 CAS-Nummer: 221313-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...