3-Chlor-5-fluorphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-5-fluorobenzeneboronic acid, 3-Chloro-5-fluorophenlyboronic acid

PC1618-1G PC1618-5G PC1618-25G
APOSPC1618-1GEA 21.9 EUR
APOSPC1618-1G APOSPC1618-5G APOSPC1618-25G
3-Chlor-5-fluorphenylboronsäure 97%
3-Chlor-5-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
Molecular Weight: 174.37 g/mol
Schmelzpunkt: 272…276 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06801741
CAS-Nummer: 328956-61-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR