3-Ethoxy-5-fluorobenzeneboronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Ethoxy-5-fluorophenyl)boronic acid, 3-Ethoxy-5-fluorophenylboronic acid, 3-Ethoxy-5-fluorobenzeneboronic acid

PC1624-1G PC1624-5G
APOSPC1624-1GEA 142 EUR
APOSPC1624-1G APOSPC1624-5G
3-Ethoxy-5-fluorobenzeneboronic acid 95%
3-Ethoxy-5-fluorobenzeneboronic acid
Formel: C₈H₁₀BFO₃
Molecular Weight: 183,97 g/mol
Schmelzpunkt: 155…158 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07363779
CAS-Nummer: 850589-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR