5-Fluoro-2-hydroxybenzeneboronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-2-hydroxyphenylboronic acid, (5-Fluoro-2-hydroxyphenyl)boronic acid, 5-Fluoro-2-hydroxybenzeneboronic acid

PC1627-5G PC1627-1G PC1627-250MG
APOSPC1627-5GEA 66.7 EUR
APOSPC1627-5G APOSPC1627-1G APOSPC1627-250MG
5-Fluoro-2-hydroxybenzeneboronic acid 97%
5-Fluoro-2-hydroxybenzeneboronic acid
Formel: C₆H₆BFO₃
Schmelzpunkt: 160…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095357
CAS-Nummer: 259209-20-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR