4-Chlor-2-fluorphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chlor-2-fluorbenzolboronsäure, (4-Chlor-2-fluorphenyl)metaborsäure, Dihydroxy(4-chlor-2-fluorphenyl)boran

PC1629-25G PC1629-5G
APOSPC1629-25GEA 116 EUR
APOSPC1629-25G APOSPC1629-5G
4-Chlor-2-fluorphenylboronsäure 97%
4-Chlor-2-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
Molecular Weight: 174,37 g/mol
Schmelzpunkt: 246…250 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02684293
CAS-Nummer: 160591-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR