2-Chlor-5-fluorphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-5-fluorobenzeneboronic acid
PC1631-1G
PC1631-25G
PC1631-5G
APOSPC1631-1GEA
21.9
EUR
APOSPC1631-1G
APOSPC1631-25G
APOSPC1631-5G
2-Chlor-5-fluorphenylboronsäure 98%
2-Chlor-5-fluorphenylboronsäure
Formel:
C₆H₅BClFO₂ Molecular Weight: 174.37 g/mol Schmelzpunkt: 78…82 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06656272 CAS-Nummer: 444666-39-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...