5-Chlor-2-fluorphenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Dihydroxy(5-chlor-2-fluorphenyl)boran, 5-Chlor-2-fluorbenzolboronsäure, (5-Chlor-2-fluorphenyl)metaborsäure
PC1635-25G
PC1635-5G
APOSPC1635-25GEA
302
EUR
APOSPC1635-25G
APOSPC1635-5G
5-Chlor-2-fluorphenylboronsäure 97%
5-Chlor-2-fluorphenylboronsäure
Formel:
C₆H₅BClFO₂ Molecular Weight: 174,37 g/mol Schmelzpunkt: 182…188 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05664225 CAS-Nummer: 352535-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...