2-Chlor-4-fluorphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC1636-25G
PC1636-5G
APOSPC1636-25GEA
84.1
EUR
APOSPC1636-25G
APOSPC1636-5G
2-Chlor-4-fluorphenylboronsäure 98%
2-Chlor-4-fluorphenylboronsäure
Formel:
C₆H₅BClFO₂ Molecular Weight: 174,37 g/mol Schmelzpunkt: 220…223 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02684295 CAS-Nummer: 313545-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...