3-Cyan-4-fluorphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Cyano-4-fluorobenzeneboronic acid

PC1637-1G PC1637-5G
APOSPC1637-1GEA 21.9 EUR
APOSPC1637-1G APOSPC1637-5G
3-Cyan-4-fluorphenylboronsäure 97%
3-Cyan-4-fluorphenylboronsäure
Formel: C₇H₅BFNO₂
Molecular Weight: 164,93 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095130
CAS-Nummer: 214210-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR